4-Benzylcarbamoyl-2-fluorobenzeneboronic acid, 97%

H55058.MDEA 72.3 EUR
ALFAH55058.MD ALFAH55058.03
4-Benzylcarbamoyl-2-fluorobenzeneboronic acid, 97%
Purity: 97%
CAS No.: 874289-36-8
Molecular Formula: C14H13BFNO3
Formula Weight: 273.07
MDL No.: MFCD20040289
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR