3-Benzylcarbamoyl-5-fluorobenzeneboronic acid, 98%

H52527.03EA 155.1 EUR
ALFAH52527.03 ALFAH52527.MD
3-Benzylcarbamoyl-5-fluorobenzeneboronic acid, 98%
Purity: 98%
CAS No.: 874219-41-7
Molecular Formula: C14H13BFNO3
Formula Weight: 273.07
Melting Point: 263-265°
MDL No.: MFCD08235058
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR