2-Fluoro-5-(3-fluoro-4-methylbenzylcarbamoyl)phenylboronic acid ≥97%

Supplier: Thermo Scientific
Warning

Synonyms: 2-Fluoro-5-(3-fluoro-4-methylbenzylcarbamoyl)benzeneboronic acid

H54230.03EA 270 EUR
H54230.03 H54230.MD
2-Fluoro-5-(3-fluoro-4-methylbenzylcarbamoyl)phenylboronic acid ≥97%
2-Fluoro-5-(3-fluoro-4-methylbenzylcarbamoyl)phenylboronic acid
Formula: C₁₅H₁₄BF₂NO₃
Storage Temperature: Ambient
MDL Number: MFCD20040159
CAS Number: 1449132-35-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR