Propargite, Supelco®

Supplier: Merck


M-Clarity™ quality level = MQ100

32051-100MGEA 71 EUR
32051-100MG
Propargite, Supelco®
Propargite
Formula: C₁₉H₂₆O₄S
MDL Number: MFCD00072448
CAS Number: 2312-35-8
EINECS: 219-006-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR