Barium-2-ethylhexanoate 20% (w/w) in 2-ethylhexanoic acid
Supplier: Thermo Scientific
44021.18EA
206
EUR
44021.18
Barium-2-ethylhexanoate 20% (w/w) in 2-ethylhexanoic acid
Barium 2-ethylhexanoate
CAS No.: 2457-01-4
Molecular Formula: C16H30BaO4
Formula Weight: 423.76
Physical Form: Liquid
Odour: Weak, characteristic
MDL No.: MFCD00058696
Formula:
C₁₆H₃₀BaO₄ MW: 423,76 g/mol |
MDL Number:
MFCD00058696 CAS Number: 2457-01-4 EINECS: 219-535-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...