2,2,4'-Tribromoacetophenone ≥97%

Supplier: Thermo Scientific
Danger

H27466.06EA 31.3 EUR
H27466.06 H27466.14
2,2,4'-Tribromoacetophenone ≥97%
2,2,4'-Tribromoacetophenone
Formula: C₈H₅Br₃O
MW: 356,84
Melting Pt: 91…94 °C
Storage Temperature: Ambient
MDL Number: MFCD00017858
CAS Number: 13195-79-4
EINECS: 236-161-0
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR