4-tert-Butylphenylboronic acid

Synonyms: 4-tert-Butylbenzeneboronic acid

MAYBAC35897.10EA 0 EUR
MAYBAC35897.10 MAYBAC35897.1 MAYBAC35897.50
4-tert-Butylphenylboronic acid
4-tert-Butylphenylboronic acid
Formula: C₁₀H₁₅BO₂
MW: 178,04
MDL Number: MFCD01009697
CAS Number: 123324-71-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR