4-Propylphenylboronic acid ≥98%

Supplier: Thermo Scientific
Warning

Synonyms: 4-n-Propylbenzeneboronic acid4-n-Propylphenylboronic acid

H52884.03EA 30.8 EUR
H52884.03 H52884.06
4-Propylphenylboronic acid ≥98%
4-Propylphenylboronic acid
Formula: C₉H₁₃BO₂
MW: 164,01
Melting Pt: 106…108 °C
Storage Temperature: Ambient
MDL Number: MFCD00859261
CAS Number: 134150-01-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR