3-Fluoro-4-(trifluoromethoxy)benzyl bromide ≥97%

Supplier: Thermo Scientific
Danger

H26275.MDEA 81.5 EUR
H26275.MD H26275.03
3-Fluoro-4-(trifluoromethoxy)benzyl bromide ≥97%
3-Fluoro-4-(trifluoromethoxy)benzyl bromide
Formula: C₈H₅BrF₄O
Storage Temperature: Ambient
MDL Number: MFCD06660205
CAS Number: 886499-04-3
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR