4-Fluoro-3-(trifluoromethoxy)benzyl bromide ≥97%

Supplier: Thermo Scientific
Danger

H26343.03EA 243 EUR
H26343.03 H26343.MD
4-Fluoro-3-(trifluoromethoxy)benzyl bromide ≥97%
4-Fluoro-3-(trifluoromethoxy)benzyl bromide
Formula: C₈H₅BrF₄O
Storage Temperature: Ambient
MDL Number: MFCD06660256
CAS Number: 86256-50-0
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR