2-Fluoro-3-(trifluoromethyl)phenylboronic acid ≥97%

Warning

Synonyms: 2-Fluoro-3-(trifluoromethyl)benzeneboronic acid

H53364.03EA 104 EUR
H53364.03 H53364.06
2-Fluoro-3-(trifluoromethyl)phenylboronic acid ≥97%
2-Fluoro-3-(trifluoromethyl)phenylboronic acid
Formula: C₇H₅BF₄O₂
MW: 207,92
Melting Pt: 56…58 °C
Storage Temperature: Ambient
MDL Number: MFCD07784336
CAS Number: 157834-21-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR