3-Fluoro-5-(trifluoromethyl)phenylboronic acid ≥97%

Warning

Synonyms: 3-Fluoro-5-(trifluoromethyl)benzeneboronic acid

H52526.MDEA 51.7 EUR
H52526.MD H52526.03
3-Fluoro-5-(trifluoromethyl)phenylboronic acid ≥97%
3-Fluoro-5-(trifluoromethyl)phenylboronic acid
Formula: C₇H₅BF₄O₂
MW: 207,92
Storage Temperature: Ambient
MDL Number: MFCD06657735
CAS Number: 159020-59-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR