4-Fluoro-2-(trifluoromethyl)phenylboronic acid ≥98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
(4-Fluoro-2-(trifluoromethyl)phenyl)boronic acid
, 4-Fluoro-2-(trifluoromethyl)benzeneboronic acid
H26144.03EA
267
EUR
H26144.03
4-Fluoro-2-(trifluoromethyl)phenylboronic acid ≥98%
4-Fluoro-2-(trifluoromethyl)phenylboronic acid
Formula:
C₇H₅BF₄O₂ MW: 207,92 g/mol Melting Pt: 183…186 °C Storage Temperature: Ambient |
MDL Number:
MFCD08062379 CAS Number: 182344-16-7 EINECS: None |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...