4-Fluoro-3-methylphenylboronic acid ≥98%

Supplier: Thermo Scientific
Warning

Synonyms: 4-Fluoro-3-methylbenzeneboronic acid

L18753.03EA 0 EUR
L18753.03
4-Fluoro-3-methylphenylboronic acid ≥98%
4-Fluoro-3-methylphenylboronic acid
Formula: C₇H₈BFO₂
MW: 153,95
Melting Pt: 208…211 °C
Storage Temperature: Ambient
MDL Number: MFCD01863527
CAS Number: 139911-27-6
EINECS: 000-000-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR