2'-Fluoro-4'-(trifluoromethyl)acetophenone ≥97%

Supplier: Thermo Scientific
Warning

B23517.06EA 163 EUR
B23517.06 B23517.03
2'-Fluoro-4'-(trifluoromethyl)acetophenone ≥97%
2'-Fluoro-4'-(trifluoromethyl)acetophenone
Formula: C₉H₆F₄O
Storage Temperature: Ambient
MDL Number: MFCD00061247
CAS Number: 122023-29-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR