4'-Fluoro-2'-(trifluoromethyl)acetophenone ≥97%

Supplier: Thermo Scientific

Warning

Synonyms: 1-(4-Fluoro-2-(trifluoromethyl)phenyl)ethanone

B20558.06EA 167 EUR
B20558.06 B20558.03
4'-Fluoro-2'-(trifluoromethyl)acetophenone ≥97%
4'-Fluoro-2'-(trifluoromethyl)acetophenone
Formula: C₉H₆F₄O
MW: 206,14
Storage Temperature: Ambient
MDL Number: MFCD00061251
CAS Number: 208173-21-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR