2,3-Difluoro-4-methylbenzylamine 97%

Supplier: Thermo Scientific
Danger

H26715.06EA 340 EUR
H26715.06 H26715.03
2,3-Difluoro-4-methylbenzylamine 97%
2,3-Difluoro-4-methylbenzylamine
Formula: C₈H₉F₂N
Boiling Pt: 42 °C (1 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD01631409
CAS Number: 261763-41-1
EINECS: None
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR