3-Ethoxy-2,6-difluorobenzylamine 97%, JRD

Supplier: Thermo Scientific

Danger

H31587.03EA 247 EUR
H31587.03 H31587.MD
3-Ethoxy-2,6-difluorobenzylamine 97%, JRD
3-Ethoxy-2,6-difluorobenzylamine
Formula: C₉H₁₁F₂NO
Storage Temperature: Ambient
MDL Number: MFCD11519371
CAS Number: 1092461-29-4
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR