2,6-Difluoro-3-methoxybenzylamine ≥97%

Supplier: Thermo Scientific

H26400.03EA 0 EUR
H26400.03
2,6-Difluoro-3-methoxybenzylamine ≥97%
2,6-Difluoro-3-methoxybenzylamine
Formula: C₈H₉F₂NO
MDL Number: MFCD04115905
CAS Number: 886498-50-6
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR