3-Fluoro-2-methoxybenzylamine 97%

Supplier: Thermo Scientific
Danger

H32293.03EA 270 EUR
H32293.03 H32293.MD
3-Fluoro-2-methoxybenzylamine 97%
3-Fluoro-2-methoxybenzylamine
Formula: C₈H₁₀FNO
Storage Temperature: Ambient
MDL Number: MFCD09025368
CAS Number: 916420-49-0
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR