3-Fluoro-4-(trifluoromethoxy)benzylamine 97%

Supplier: Thermo Scientific
Danger

H26334.03EA 113 EUR
H26334.03 H26334.MD
3-Fluoro-4-(trifluoromethoxy)benzylamine 97%
3-Fluoro-4-(trifluoromethoxy)benzylamine
Formula: C₈H₇F₄NO
Storage Temperature: Ambient
MDL Number: MFCD06660207
CAS Number: 886499-13-4
EINECS: None
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR