4-Fluoro-2-(trifluoromethoxy)benzylamine 97%
|
Danger
|
H33277.MDEA
38.3
EUR
H33277.MD
H33277.03
4-Fluoro-2-(trifluoromethoxy)benzylamine 97%
4-Fluoro-2-(trifluoromethoxy)benzylamine
Formula:
C₈H₇F₄NO MW: 209,14 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD19687190 CAS Number: 1241828-99-8 UN: 3267 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...