4-Fluoro-3-(trifluoromethoxy)benzylamine 97%

Supplier: Thermo Scientific

Danger

H26858.MDEA 33.9 EUR
H26858.MD H26858.03
4-Fluoro-3-(trifluoromethoxy)benzylamine 97%
4-Fluoro-3-(trifluoromethoxy)benzylamine
Formula: C₈H₇F₄NO
Storage Temperature: Ambient
MDL Number: MFCD06660254
CAS Number: 886501-20-8
EINECS: None
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR