Methyl perfluorododecanoate 96%, rods

L16809.06EA 45.4 EUR
L16809.06 L16809.14
Methyl perfluorododecanoate 96%, rods
Methyl perfluorododecanoate
Formula: C₁₃H₃F₂₃O₂
Melting Pt: 54…56 °C
Density: 1,660
Storage Temperature: Ambient
MDL Number: MFCD00236616
CAS Number: 56554-52-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR