2,4-Dichloro-3-ethyl-6-nitrophenol ≥98%

Supplier: Thermo Scientific
Danger

B22261.09EA 98.2 EUR
B22261.09 B22261.88
2,4-Dichloro-3-ethyl-6-nitrophenol ≥98%
2,4-Dichloro-3-ethyl-6-nitrophenol
Formula: C₈H₇Cl₂NO₃
Melting Pt: 49…51 °C
Storage Temperature: Ambient
MDL Number: MFCD00270764
CAS Number: 99817-36-4
EINECS: 420-740-1
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR