2-Chloro-4-(trifluoromethyl)benzyl bromide ≥97%
|
Danger
|
H33311.MDEA
94.1
EUR
H33311.MD
H33311.03
2-Chloro-4-(trifluoromethyl)benzyl bromide ≥97%
2-Chloro-4-(trifluoromethyl)benzyl bromide
Formula:
C₈H₅BrClF₃ MW: 273,48 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD14155638 CAS Number: 279252-26-5 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...