2-Methyl-5-(trifluoromethyl)benzyl bromide ≥97%

Supplier: Thermo Scientific

Danger

H26554.MDEA 40.3 EUR
H26554.MD H26554.03
2-Methyl-5-(trifluoromethyl)benzyl bromide ≥97%
2-Methyl-5-(trifluoromethyl)benzyl bromide
Formula: C₉H₈BrF₃
Storage Temperature: Ambient
MDL Number: MFCD01631609
CAS Number: 261952-17-4
UN: 3265
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR