4-Methoxy-2-(trifluoromethyl)benzyl bromide 97%

Supplier: Thermo Scientific
Danger

H31871.06EA 362 EUR
H31871.06 H31871.03
4-Methoxy-2-(trifluoromethyl)benzyl bromide 97%
4-Methoxy-2-(trifluoromethyl)benzyl bromide
Formula: C₉H₈BrF₃O
Storage Temperature: Ambient
MDL Number: MFCD09025406
CAS Number: 916420-86-5
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR