Ytterbium(III) perchlorate 50% (w/w) in aqueous solution, Reagent Grade
Supplier: Thermo Scientific
|
Danger
|
40577.NAEA
42.6
EUR
40577.NA
40577.NC
Ytterbium(III) perchlorate 50% (w/w) in aqueous solution, Reagent Grade
Ytterbium (III) perchlorate
Formula:
Cl₃O₁₂Yb Storage Temperature: Ambient |
MDL Number:
MFCD00049612 EINECS: 236-823-9 UN: 3211 ADR: 5.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...