Tetrahydro-4H-thiopyran-4-one

Synonyms: Tetrahydrothiopyran-4-one

MAYBRF03869DAEA 0 EUR
MAYBRF03869DA
Tetrahydro-4H-thiopyran-4-one
Tetrahydro-4H-thiopyran-4-one
Formula: C₅H₈OS
MW: 116,18
MDL Number: MFCD00006660
CAS Number: 1072-72-6
EINECS: 214-015-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR