2'-Methyl-5'-(trifluoromethyl)acetophenone ≥97%
|
Warning
|
H31819.06EA
316
EUR
H31819.06
H31819.03
2'-Methyl-5'-(trifluoromethyl)acetophenone ≥97%
2'-Methyl-5'-(trifluoromethyl)acetophenone
Formula:
C₁₀H₉F₃O MW: 202,17 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09832290 CAS Number: 725743-50-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...