4'-Methyl-3'-(trifluoromethyl)acetophenone ≥97%

Supplier: Thermo Scientific

Warning

H32226.MDEA 53.1 EUR
H32226.MD H32226.03
4'-Methyl-3'-(trifluoromethyl)acetophenone ≥97%
4'-Methyl-3'-(trifluoromethyl)acetophenone
Formula: C₁₀H₉F₃O
Storage Temperature: Ambient
MDL Number: MFCD08532499
CAS Number: 851262-98-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR