1,1,1,5,5,6,6,7,7,7-Decafluoro-2,4-heptanedione ≥97%

Supplier: Thermo Scientific

Danger

L09724.06EA 110 EUR
L09724.06 L09724.14 L09724.03
1,1,1,5,5,6,6,7,7,7-Decafluoro-2,4-heptanedione ≥97%
1,1,1,5,5,6,6,7,7,7-Decafluoro-2,4-heptanedione
Formula: C₇H₂F₁₀O₂
MW: 308,08
Boiling Pt: 98…100 °C
Storage Temperature: Ambient
MDL Number: MFCD00040961
CAS Number: 20583-66-8
UN: 3265
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR