Perfluoropelargonic acid ≥97%
|
Danger
|
Synonyms:
Perfluorononanoic acid
A13980.06EA
69.2
EUR
A13980.06
A13980.14
A13980.22
Perfluoropelargonic acid ≥97%
Perfluoropelargonic acid
Formula:
C₉HF₁₇O₂ MW: 464,07 g/mol Boiling Pt: 218 °C Melting Pt: 63…68 °C Storage Temperature: Ambient |
MDL Number:
MFCD00039605 CAS Number: 375-95-1 EINECS: 206-801-3 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...