4-Fluoro-N-n-propylbenzylamine, 97%

H56563.03EA 289.7 EUR
ALFAH56563.03 ALFAH56563.MD
4-Fluoro-N-n-propylbenzylamine, 97%
Purity: 97%
CAS No.: 741698-80-6
Molecular Formula: C10H14FN
Formula Weight: 167.22
UN No.: 3259
MDL No.: MFCD04489393
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR