4'-Hydroxyoctanophenone ≥99%
Supplier: Thermo Scientific
|
Warning
|
L11662.03EA
47.8
EUR
L11662.03
L11662.06
4'-Hydroxyoctanophenone ≥99%
4'-Hydroxyoctanophenone
Formula:
C₁₄H₂₀O₂ MW: 220,31 g/mol Melting Pt: 62…65 °C Storage Temperature: Ambient |
MDL Number:
MFCD00082694 CAS Number: 2589-73-3 EINECS: 219-979-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...