Potassium hydrogen diiodate 99.85%, Extra Pure for analysis
|
Danger
|
Synonyms:
Potassium bi-iodate
ACRO211475000EA
404
EUR
ACRO211475000
ACRO211471000
Potassium hydrogen diiodate 99.85%, Extra Pure for analysis
Potassium hydrogen diiodate
Formula:
HI₂KO₆ MW: 389,91 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00011400 CAS Number: 13455-24-8 EINECS: 236-650-9 UN: 3085 ADR: 5.1,II |
Specification Test Results
Appearance | White to almost white Powder or crystals |
Titration Iodometric | 99.80 to 100.20 % |
Sulfate (SO4) | ≤50 ppm |
Cl, ClO3, Br, BrO3 (as Cl) | ≤200 ppm |
Calcium (Ca) | ≤20 ppm |
Iron (Fe) | ≤20 ppm |
Lead (Pb) | ≤5 ppm |
Sodium (Na) | ≤50 ppm |
Total nitrogen (as N) | ≤20 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...