Potassium hydrogen diiodate ≥99.8% (RT), Puriss. p.a., Fluka™

Supplier: Honeywell Chemicals
Danger

Synonyms: Potassium bi-iodate

60350-100GEA 89.17 EUR
60350-100G
Potassium hydrogen diiodate ≥99.8% (RT), Puriss. p.a., Fluka™
Potassium hydrogen diiodate
Formula: HI₂KO₆
MW: 389,91
Storage Temperature: Ambient
MDL Number: MFCD00011400
CAS Number: 13455-24-8
EINECS: 236-650-9
UN: 3085
ADR: 5.1,II

Order Now

Specification Test Results

Assay 99.8 - 100.2 %
Calcium (Ca) max. 20 ppm
Cadmium (Cd) max. 5 ppm
Cobalt (Co) max. 5 ppm
Chromium (Cr) max. 5 ppm
Copper (Cu) max. 5 ppm
Iron (Fe) max. 20 ppm
Magnesium (Mg) max. 5 ppm
Manganese (Mn) max. 5 ppm
Sodium (Na) max. 50 ppm
Nickel (Ni) max. 5 ppm
Lead (Pb) max. 5 ppm
Zinc (Zn) max. 20 ppm
Total nitrogen (as N) max. 20 ppm
Sulfate (SO₄) max. 50 ppm
Chlorate, Chloride, Bromide, Bromate (as Cl) max. 200 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR