4-Isopropoxy-2-methylphenylboronic acid ≥98%

Supplier: Thermo Scientific
Warning

Synonyms: 4-Isopropoxy-2-methylbenzeneboronic acid

H53132.MDEA 0 EUR
H53132.MD
4-Isopropoxy-2-methylphenylboronic acid ≥98%
4-Isopropoxy-2-methylphenylboronic acid
Formula: C₁₀H₁₅BO₃
MW: 194,04
Melting Pt: 112…116 °C
Storage Temperature: Ambient
MDL Number: MFCD07363788
CAS Number: 871126-21-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR