3-Bromo-5-(trifluoromethyl)phenylboronic acid ≥98%

Supplier: Thermo Scientific
Warning

Synonyms: 3-Bromo-5-(trifluoromethyl)benzeneboronic acid

H52898.MDEA 0 EUR
H52898.MD
3-Bromo-5-(trifluoromethyl)phenylboronic acid ≥98%
3-Bromo-5-(trifluoromethyl)phenylboronic acid
Formula: C₇H₅BBrF₃O₂
MW: 268,82
Melting Pt: 177…180 °C
Storage Temperature: Ambient
MDL Number: MFCD08436051
CAS Number: 913835-64-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR