4'-Methoxy-3'-nitroacetophenone ≥98%

Supplier: Thermo Scientific

L04149.09EA 69.5 EUR
L04149.09 L04149.18
4'-Methoxy-3'-nitroacetophenone ≥98%
4'-Methoxy-3'-nitroacetophenone
Formula: C₉H₉NO₄
MW: 195,18
Melting Pt: 97…99 °C
Storage Temperature: Ambient
MDL Number: MFCD00017023
CAS Number: 6277-38-9
EINECS: 228-476-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR