Ethyl-6-(chloroformyl)hexanoate 95%

Supplier: Thermo Scientific

Warning

H50714.03EA 187 EUR
H50714.03 H50714.06
Ethyl-6-(chloroformyl)hexanoate 95%
Ethyl 6-(chloroformyl)hexanoate
Formula: C₉H₁₅ClO₃
Storage Temperature: Ambient
MDL Number: MFCD00671507
CAS Number: 14794-32-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR