2-Methyl-3-(trifluoromethyl)benzylamine 97%

Supplier: Thermo Scientific
Danger

H31552.MDEA 49.5 EUR
H31552.MD H31552.03
2-Methyl-3-(trifluoromethyl)benzylamine 97%
2-Methyl-3-(trifluoromethyl)benzylamine
Formula: C₉H₁₀F₃N
Storage Temperature: Ambient
MDL Number: MFCD06213551
CAS Number: 771572-43-1
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR