3-Methyl-5-(trifluoromethyl)benzylamine 97%
|
Danger
|
H33921.MDEA
38.3
EUR
H33921.MD
H33921.03
3-Methyl-5-(trifluoromethyl)benzylamine 97%
3-Methyl-5-(trifluoromethyl)benzylamine
Formula:
C₉H₁₀F₃N MW: 189,18 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06213553 CAS Number: 292151-97-4 UN: 3267 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...