Diphenylamine, BAKER ANALYZED® ACS, J.T. Baker®

Supplier: Avantor
Danger

Synonyms: N-PhenylbenzeneamineN-Phenylaniline

1944-04EA 0 EUR
1944-04 1944-01
Diphenylamine, BAKER ANALYZED® ACS, J.T. Baker®
Diphenylamine

Crystals. Lot analysis on label.

Formula: (C₆H₅)₂NH
MW: 169,23 g/mol
Boiling Pt: 302 °C (1013 hPa)
Melting Pt: 52 °C
Density: 1,165 g/cm³ (20 °C)
Flash Pt: 153 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00003014
CAS Number: 122-39-4
EINECS: 204-539-4
ADR: 9,III
Merck Index: 13,03349
https://ie.vwr.com/stibo/web/std.lang.all/76/36/2297636.jpg

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Melting Point 52.5 - 54.0 °C
Solubility in Alcohol Passes Test
Residue after Ignition ≤0.03 %
Nitrate (NO₃) Passes Test
Sensitivity for Nitrate Passes Test
Appearance (white to cream-white crystal line powder)
Meets Reagent Specifications for testing USP/NF monographs

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR