Polyvinylpyrrolidone, M.W. ∼ 360,000 (K90)

Supplier: Thermo Scientific

Synonyms: PolyvidonePolyvinylpyrrolidonPVP

J61381.36EA 115 EUR
J61381.36 J61381.A1 J61381.30
Polyvinylpyrrolidone, M.W. ∼ 360,000 (K90)
Polyvinylpyrrolidone

A cryoprotectant, also useful in plant cell culture research

Formula: (C₆H₉NO)n
Storage Temperature: Ambient
MDL Number: MFCD01076626
CAS Number: 9003-39-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR