1,5-Diphenylcarbazide, powder, BAKER ANALYZED® ACS, J.T.Baker®
|
Warning
|
Synonyms:
sym. Diphenylcarbazide
, sym-Diphenylcarbazide
, N'',N'''-Diphenylcarbonohydrazide
, 1,5-Diphenylcarbohydrazide
, Diphenylcarbazide
, 1,5-Diphenylcarbonohydrazide
K620-03EA
154.12
EUR
K620-03
1,5-Diphenylcarbazide, powder, BAKER ANALYZED® ACS, J.T.Baker®
1,5-Diphenylcarbazide
Powder.
Formula:
C₁₃H₁₄N₄O MW: 242,28 g/mol Boiling Pt: 454 °C (1013 hPa) Melting Pt: 170…172 °C Density: 1,31 g/cm³ (20 °C) |
MDL Number:
MFCD00003013 CAS Number: 140-22-7 EINECS: 205-403-7 Merck Index: 13,03353 |
Specification Test Results
Meets ACS Reagent Chemical Requirements | |
For Laboratory,Research,or Manufacturing Use | |
Melting Point | 173 - 176 °C |
Residue after Ignition | ≤0.050 % |
Solubility in Aqueous Acetone | Passes Test |
Sensitivity for Chromate | Passes Test |
Meets Reagent Specifications for testing USP/NF monographs |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...