beta-Alanine, Cell Culture Reagent

J63435.A1EA 210.2 EUR
ALFAJ63435.A1 ALFAJ63435.36 ALFAJ63435.30
beta-Alanine, Cell Culture Reagent
CAS No.: 107-95-9
Molecular Formula: C3H7NO2
Formula Weight: 89.09
Physical Form: Powder
Density: 1.437
Appearance: White
Odour: Odourless
Melting Point: ca 205° dec.
Beilstein Reference: 906793
Merck Reference: 14,205
MDL No.: MFCD00008200
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR