Formic acid 90%, BAKER ANALYZED®, J.T.Baker®
|
Danger
|
Synonyms:
Methanoic acid
0129-01EA
196.55
EUR
0129-01
0129-05
Formic acid 90%, BAKER ANALYZED®, J.T.Baker®
Formic acid
Polymer characterization solvent.
Formula:
HCO₂H MW: 46,03 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00003297 CAS Number: 64-18-6 EINECS: 200-579-1 UN: 1779 ADR: 8, 3,II |
Specification Test Results
For Laboratory,Research,or Manufacturing Use | |
Assay (HCOOH) | 89.8 - 91.5 % |
Clarity | Passes Test |
Color (APHA) | ≤10 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...