Trichloroacetic acid (TCA) ≥99.0% ACS

Supplier: VWR Chemicals
Danger

Synonyms: TCA

0133-100GEA 0 EUR
0133-100G 0133-500G
Trichloroacetic acid (TCA) ≥99.0% ACS
Trichloroacetic acid (TCA)
Formula: Cl₃CCOOH
MW: 163,39 g/mol
Boiling Pt: 196 °C (1013 hPa)
Melting Pt: 54…56 °C
Density: 1,63 g/cm³ (20 °C)
Flash Pt: 113 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00004177
CAS Number: 76-03-9
EINECS: 200-927-2
Merck Index: 12,09756
https://ie.vwr.com/stibo/web/std.lang.all/74/52/727452.jpg

Order Now

Specification Test Results

Chloride <0.002%
Clarity of Solution Pass
Heavy Metals ≤0.002%
Insoluble Matter ≤0.01%
Iron ≤0.001%
Nitrate ≤0.002%
Phosphate ≤5 ppm
Purity ≥99%
Residue after Ignition ≤0.03%
Substances Darkened by H2SO4 Pass
Sulphate ≤0.02%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR